Merck
Pharmacological properties of alpha-mangostin, a novel histamine H1 receptor antagonist.
European journal of pharmacology (1996-10-31)
N Chairungsrilerd, K Furukawa, T Ohta, S Nozoe, Y Ohizumi
RESUMEN
In the isolated rabbit thoracic aorta and guinea-pig trachea, alpha-mangostin inhibited histamine-induced contractions in a concentration-dependent manner in the presence or absence of cimetidine, a histamine H2 receptor antagonist. But KCl-, phenylephrine- or carbachol-induced contractions were not affected by alpha-mangostin. The concentration-contractile response curve for histamine was shifted to the right in a parallel manner by alpha-mangostin. In the presence of chlorpheniramine, a histamine H1 receptor antagonist, alpha-mangostin did not affect the relaxation of the rabbit aorta induced by histamine. In the guinea-pig trachea, alpha-mangostin had no effect on the relaxation induced by dimaprit, a histamine H2 receptor agonist. alpha-Mangostin caused a concentration-dependent inhibition of the binding of [3H]mepyramine, a specific histamine H1 receptor antagonist to rat aortic smooth muscle cells. Kinetic analysis of [3H]mepyramine binding indicated the competitive inhibition by alpha-mangostin. These results suggest that alpha-mangostin is a novel competitive histamine H1 receptor antagonist in smooth muscle cells.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
α-Mangostin, ≥98% (HPLC)