Merck
Increased levels of conjugated bile acids are associated with human bile reflux gastritis.
Scientific reports (2020-07-16)
Aihua Zhao, Shouli Wang, Wenlian Chen, Xiaojiao Zheng, Fengjie Huang, Xiaolong Han, Kun Ge, Cynthia Rajani, Yanxia Huang, Herbert Yu, Jinshui Zhu, Wei Jia
RESUMEN
Bile acids (BAs) play essential roles in facilitating lipid digestion and absorption in the intestine. Gastric BAs were attributed to abnormal refluxing from duodenal compartments and correlated with the occurrence of gastric inflammation and carcinogenesis. However, the differences in gastric BAs between physiologically compromised and healthy individuals have not been fully investigated. In this study, gastric juice was collected from patients clinically diagnosed as gastritis with/without bile reflux and healthy subjects for BA profiles measurements. As a result, we found that the conjugated BAs became prominent components in bile reflux juice, whereas almost equal amounts of conjugated and unconjugated BAs existed in non-bile reflux and healthy juice. To investigate whether gastric BA changes were regulated by hepatic BA synthesis, C57BL/6J mice were intervened with GW4064/resin to decrease/increase hepatic BA synthesis. The results revealed that changes of gastric BAs were coordinated with hepatic BA changes. Additionally, gastric BAs were detected in several healthy mammals, in which there were no obvious differences between the conjugated and unconjugated BAs. Pigs were an exception. Thus, increased levels of conjugated BAs are associated with human bile reflux gastritis. Gastric conjugated BAs could become a panel of biomarkers to facilitate diagnosis of pathological bile reflux.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)
Sigma-Aldrich
Lithocholic acid-2,2,4,4-d4, 98 atom % D