Merck
A cryptic RNA-binding domain mediates Syncrip recognition and exosomal partitioning of miRNA targets.
Nature communications (2018-02-28)
Fruzsina Hobor, Andre Dallmann, Neil J Ball, Carla Cicchini, Cecilia Battistelli, Roksana W Ogrodowicz, Evangelos Christodoulou, Stephen R Martin, Alfredo Castello, Marco Tripodi, Ian A Taylor, Andres Ramos
RESUMEN
Exosomal miRNA transfer is a mechanism for cell-cell communication that is important in the immune response, in the functioning of the nervous system and in cancer. Syncrip/hnRNPQ is a highly conserved RNA-binding protein that mediates the exosomal partition of a set of miRNAs. Here, we report that Syncrip's amino-terminal domain, which was previously thought to mediate protein-protein interactions, is a cryptic, conserved and sequence-specific RNA-binding domain, designated NURR (N-terminal unit for RNA recognition). The NURR domain mediates the specific recognition of a short hEXO sequence defining Syncrip exosomal miRNA targets, and is coupled by a non-canonical structural element to Syncrip's RRM domains to achieve high-affinity miRNA binding. As a consequence, Syncrip-mediated selection of the target miRNAs implies both recognition of the hEXO sequence by the NURR domain and binding of the RRM domains 5' to this sequence. This structural arrangement enables Syncrip-mediated selection of miRNAs with different seed sequences.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-hnRNP Q Antibody, clone 18E4, clone 18E4, from mouse