Merck
Salt-induced swelling transitions of a lamellar amphiphile-Polyelectrolyte complex.
The Journal of chemical physics (2019-03-10)
Meera Thomas, K Swamynathan, V A Raghunathan
RESUMEN
We report salt-induced swelling transitions of a lamellar complex of the anionic polyelectrolyte, poly(acrylic acid sodium salt) (PAANa), and the cationic amphiphile, didodecyldimethylammonium chloride (DDAC). Increasing the concentration of NaCl in the solution is found to lead to a collapsed → swollen → collapsed transition of the complex. The swelling transition is driven by an abrupt increase in PAANa adsorption on DDAC bilayers above a threshold salt concentration. The lamellar periodicity of the swollen phase is not determined by the thickness of the adsorption layer, and additional mechanisms have to be invoked to understand the extent of its swelling. The swelling transition is not observed for the highest molecular weight of PAANa used, but a gradual transformation between the two collapsed structures is seen on increasing the salt concentration. The polyelectrolyte chains desorb from the bilayers at a very high salt concentration, in a process similar to the well-known destabilization of complexes of oppositely charged polyelectrolytes. However, unlike the PAANa chains, the polymer-free bilayers do not disperse uniformly in the solution. Instead, they form a collapsed lamellar stack containing very little water due to the van der Waals attraction between them. The occurrence of an abrupt swelling transition at intermediate salt concentrations in this system contrasts sharply with the gradual swelling reported in other complexes with increasing salt concentration. Furthermore, this behavior does not seem to have been anticipated by theories of complexation of oppositely charged macroions. More experiments are required for a clear understanding of the interactions stabilizing the different phases observed in this system.