Merck
A strong integrated strength and toughness artificial nacre based on dopamine cross-linked graphene oxide.
ACS nano (2014-08-12)
Wei Cui, Mingzhu Li, Jiyang Liu, Ben Wang, Chuck Zhang, Lei Jiang, Qunfeng Cheng
RESUMEN
Demands of the strong integrated materials have substantially increased across various industries. Inspired by the relationship of excellent integration of mechanical properties and hierarchical nano/microscale structure of the natural nacre, we have developed a strategy for fabricating the strong integrated artificial nacre based on graphene oxide (GO) sheets by dopamine cross-linking via evaporation-induced assembly process. The tensile strength and toughness simultaneously show 1.5 and 2 times higher than that of natural nacre. Meanwhile, the artificial nacre shows high electrical conductivity. This type of strong integrated artificial nacre has great potential applications in aerospace, flexible supercapacitor electrodes, artificial muscle, and tissue engineering.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Carboxylated reduced graphene oxide