Merck
Regulation of the Epstein-Barr viral immediate early BRLF1 promoter through a distal NF1 site.
Archives of virology (1998-12-18)
G Glaser, M Vogel, H Wolf, H H Niller
RESUMEN
The immediate early BRLF1 and BZLF1 promoters of Epstein-Barr virus are crucial for triggering the replicative cycle of the virus. To better understand the cell type dependence of the lytic cycle we conducted an analysis of the BRLF1-promoter in the epithelial cell line HeLa and the lymphoid cell line IM9. To analyze promoter activities, transient transfections with 5'-deletions of the BRLF1-promoter in front of luciferase as reporter gene were conducted. Besides the already known cis-acting elements of the promoter close to the TATA-box, more distal elements were located and functionally tested. A nuclear factor 1 consensus site was found to act positively in HeLa cells, but did not in lymphoid IM9 cells. The NF1 site was shown to bind protein by electrophoretic mobility shift assays, antibody-supershifts and in vitro footprinting. Thus, a protein belonging to the nuclear factor 1 family of proteins was identified as additional cellular trans-acting factor for the BRLF1-promoter besides the already described factors Sp1, Zta and Zif268.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Heparin−Agarose, Type I, saline suspension