Merck
The role of α2,3-linked sialylation on clear cell type epithelial ovarian cancer.
Taiwanese journal of obstetrics & gynecology (2018-04-21)
Pi-Lin Sung, Kuo-Chang Wen, Huann-Cheng Horng, Chia-Ming Chang, Yi-Jen Chen, Wen-Ling Lee, Peng-Hui Wang
RESUMEN
Our previous study has shown that high expression of α2,3-sialytransferase type I was associated with advanced stage serous type epithelial ovarian cancer (EOC). The aim of the current study further attempts to evaluate the altered α 2,3-sialylation on the behavior of clear cell type EOC (C-EOC). Immunohistochemistry staining, bioinformatics analysis and tissue array were used to disclose the clinical significance of over α2,3-sialylation in C-EOC. An α2,3 sialylation inhibitor, soyasaponin I (SsaI) was used to investigate the behavior change of the C-EOC cell line. We reconfirmed that α2,3-sialylation, instead of α2,6- sialylation, was associated with late-stage C-EOC. Soyasaponin I could inhibit α2,3-sialylation of C-EOC cell lines and increase E-cadherin expression with subsequently suppressing migration of C-EOC cells. The current study demonstrated the important role of α2,3-linked sialylation in C-EOC and targeting of α2,3-linked sialylation might offer as a potential therapeutic strategy in the future.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Soyasaponin I