HPLC Analysis of Diet Soda Spiked with Neotame on Ascentis® Express C18
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Express C18, 3 cm x 4.6 mm I.D., 2.7 μm particles (53818-U)
mobile phase
[A] 100 mM ammonium acetate, pH 5.6 (titrated with acetic acid); [B] water; [C] acetonitrile
gradient
20% A constant; 75 to 20% B, 5 to 60% C in 1 min; held at 20% B, 60% C for 0.1 min
flow rate
3 mL/min
pressure
3191 psi (220 bar)
column temp.
40 °C
detector
UV, 214 nm
injection
1 μL
sample
neotame 350 μg/mL in diet cola
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of common drink additives used in diet soda.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany