Merck
  • Measurement of volatile organic compounds by the US Environmental Protection Agency Compendium Method TO-17. Evaluation of performance criteria.
Measurement of volatile organic compounds by the US Environmental Protection Agency Compendium Method TO-17. Evaluation of performance criteria.
Journal of chromatography. A (1998-08-11)
W A McClenny, M Colón
ABSTRACT
An evaluation of performance criteria for US Environmental Protection Agency Compendium Method TO-17 for monitoring volatile organic compounds (VOCs) in air has been accomplished. The method is a solid adsorbent-based sampling and analytical procedure including performance criteria for four merit parameters. These are: (1) the method detection limit (MDL); (2) the method precision; (3) the agreement between two samples taken at different flow-rates over the same time period [a distributed volume pair (DVP)]; and (4) audit accuracy. A two-adsorbent tube packing of Carbotrap and Carboxen 1000 (Supelco) was tested. Synthetic mixtures containing 41 compounds at concentrations of 10 ppb (v/v) or 2 ppb (v/v) each in humidified zero air, and indoor air from a personal residence, were sampled for 1-h periods during which 1-l and 4-l samples were obtained simultaneously. For synthetic samples, the MDL was determined to be < or = 0.5 ppb (v/v) for 29 of the 41 compounds examined for samples at 47% relative humidity (RH), and for 27 of 41 compounds at 85% RH. The method precision at both 2 and 10 ppb (v/v) was < or = 20% for 90+ % of the samples using five sampling runs and the DVP samples were within 25% for 80% of the samples. For the indoor samples, 26 target compounds were tentatively identified and 12 unknowns were detected; all but 10% of these compounds met the DVP criteria.
Product Description
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Supelco
Carbotrap® 349, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Supelco
Carbotrap® 300, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
Supelco
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 563, 20-45 mesh, bottle of 10 g
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® X, 20-40 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® B, 20-40 mesh, bottle of 10 g
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® C, 20-40 mesh, bottle of 10 g
Supelco
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 1/4 in. × 5 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm