Merck
  • Heart-on-a-Chip: An Investigation of the Influence of Static and Perfusion Conditions on Cardiac (H9C2) Cell Proliferation, Morphology, and Alignment.
Heart-on-a-Chip: An Investigation of the Influence of Static and Perfusion Conditions on Cardiac (H9C2) Cell Proliferation, Morphology, and Alignment.
SLAS technology (2017-04-22)
Anna Kobuszewska, Ewelina Tomecka, Kamil Zukowski, Elzbieta Jastrzebska, Michal Chudy, Artur Dybko, Philippe Renaud, Zbigniew Brzozka
ABSTRACT
Lab-on-a-chip systems are increasingly used as tools for cultures and investigation of cardiac cells. In this article, we present how the geometry of microsystems and microenvironmental conditions (static and perfusion) influence the proliferation, morphology, and alignment of cardiac cells (rat cardiomyoblasts-H9C2). Additionally, studies of cell growth after incubation with verapamil hydrochloride were performed. For this purpose, poly(dimethylsiloxane) (PDMS)/glass microfluidic systems with three different geometries of microchambers (a circular chamber, a longitudinal channel, and three parallel microchannels separated by two rows of micropillars) were prepared. It was found that static conditions did not enhance the growth of H9C2 cells in the microsystems. On the contrary, perfusion conditions had an influence on division, morphology, and the arrangement of the cells. The highest number of cells, their parallel orientation, and their elongated morphology were obtained in the longitudinal microchannel. It showed that this kind of microsystem can be used to understand processes in heart tissue in detail and to test newly developed compounds applied in the treatment of cardiac diseases.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and sodium bicarbonate, without L-glutamine and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.