Merck
  • Identification of a Potent Phosphoinositide 3-Kinase Pan Inhibitor Displaying a Strategic Carboxylic Acid Group and Development of Its Prodrugs.
Identification of a Potent Phosphoinositide 3-Kinase Pan Inhibitor Displaying a Strategic Carboxylic Acid Group and Development of Its Prodrugs.
ChemMedChem (2017-09-01)
Tracey Pirali, Elisa Ciraolo, Silvio Aprile, Alberto Massarotti, Alex Berndt, Alessia Griglio, Marta Serafini, Valentina Mercalli, Clarissa Landoni, Carlo Cosimo Campa, Jean Piero Margaria, Rangel L Silva, Giorgio Grosa, Giovanni Sorba, Roger Williams, Emilio Hirsch, Gian Cesare Tron
ABSTRACT
Activation of the phosphoinositide 3-kinase (PI3K) pathway is a key signaling event in cancer, inflammation, and other proliferative diseases. PI3K inhibitors are already approved for some specific clinical indications, but their systemic on-target toxicity limits their larger use. In particular, whereas toxicity is tolerable in acute treatment of life-threatening diseases, this is less acceptable in chronic conditions. In the past, the strategy to overcome this drawback was to block selected isoforms mainly expressed in leukocytes, but redundancy within the PI3K family members challenges the effectiveness of this approach. On the other hand, decreasing exposure to selected target cells represents a so-far unexplored alternative to circumvent systemic toxicity. In this manuscript, we describe the generation of a library of triazolylquinolones and the development of the first prodrug pan-PI3K inhibitor.
Product Description
SAFC
Insulin, Human Recombinant, for research or for further manufacturing use, dry powder