Merck
  • Transient fusion ensures granule replenishment to enable repeated release after IgE-mediated mast cell degranulation.
Transient fusion ensures granule replenishment to enable repeated release after IgE-mediated mast cell degranulation.
Journal of cell science (2016-11-03)
Santiago Balseiro-Gomez, Juan A Flores, Jorge Acosta, M Pilar Ramirez-Ponce, Eva Ales
ABSTRACT
To ensure normal immune function, mast cells employ different pathways to release mediators. Here, we report a thus far unknown capacity of mast cells to recycle and reuse secretory granules after an antigen-evoked degranulation process under physiological conditions; this phenomenon involves the existence of a recycling secretory granule pool that is available for release in a short time scale. Rapid endocytic modes contributed to the recycling of ∼60% of the total secretory granule population, which involved kiss-and-run and cavicapture mechanisms, causing retention of the intragranular matrix. We found the presence of normal-size granules and giant actomyosin- and dynamin-dependent granules, which were characterized by large quantal content. These large structures allowed the recovered mast cells to release a large amount of 5-HT, compensating for the decrease in the number of exocytosed secretory granules. This work uncovers a new physiological role of the exo-endocytosis cycle in the immunological plasticity of mast cells and reveals a new property of their biological secretion.
Sigma-Aldrich
SynaptoGreen C4, ≥95% (HPLC), solid
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder