Merck
  • Bismuth absorption from 205Bi-labelled pharmaceutical bismuth compounds used in the treatment of peptic ulcer disease.
Bismuth absorption from 205Bi-labelled pharmaceutical bismuth compounds used in the treatment of peptic ulcer disease.
Scandinavian journal of gastroenterology (1992-04-01)
B Dresow, R Fischer, E E Gabbe, J Wendel, H C Heinrich
ABSTRACT
The absorption of bismuth from five 205Bi-labelled pharmaceutically used bismuth compounds was studied in man. From single oral doses of all compounds under investigation only less than 0.1% bismuth was absorbed and excreted with the urine. A significantly higher absorption was observed from the colloidal bismuth subcitrate (0.042% of the dose) and the basic bismuth gallate (0.038%) than from the basic bismuth salicylate, nitrate, and aluminate (0.005-0.002%). No retention of bismuth in the whole body was found from the single dose experiment. The biologic fast-term half-lives of absorbed bismuth were calculated to be 0.12 and 1.5 days.
Sigma-Aldrich
Bismuth(III) gallate basic hydrate, ≥98.0% (calc. on dry substance, KT)