Merck
  • Adsorption of naphthalene onto a high-surface-area carbon from waste ion exchange resin.
Adsorption of naphthalene onto a high-surface-area carbon from waste ion exchange resin.
Journal of environmental sciences (China) (2013-04-17)
Qianqian Shi, Aimin Li, Zhaolian Zhu, Bing Liu
ABSTRACT
A high-surface-area carbon (KC-1) was prepared from waste polystyrene-based ion exchange resin by KOH activation and used for naphthalene adsorption. The carbon exhibited a good hydrophobic nature with developed porous structure, favoring the adsorption of organic compounds. The Brunauer-Emmett-Teller surface area and total pore volume of KC-1 were 3442.2 and 1.68 cm3/g, respectively, which can be compared with those of KOH-activated carbons prepared from other precursors. Batch experiments were carried out to investigate the adsorption of naphthalene onto KC-1. The equilibrium data were analyzed by the Langmuir, Freundlich, and Polanyi-Manes isotherms and agreed with the Polanyi-Manes Model. The adsorption of naphthalene depended greatly on the porosity of the carbon, and the dispersive interactions between naphthalene and carbon could be relatively weak. The pH variation in aqueous solution had little effect on the adsorption process. The equilibrium time for 0.04 g/L of carbon dose was around 5 hr. Different models were used to evaluate the kinetic data and the pseudo second-order model was suitable to describe the kinetic process of naphthalene adsorption onto KC-1. Regeneration of spent carbon could be carried out effectively by alcohol treatment. The results indicated that KC-1 was a promising adsorbent for the removal of polycyclic aromatic hydrocarbons from aqueous solutions.
Product Description
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
Naphthalene, 99%
Supelco
Naphthalene, analytical standard
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Melting point standard 79-81°C, analytical standard