Merck
  • Aerobic oxidative coupling of arenes and olefins through a biomimetic approach.
Aerobic oxidative coupling of arenes and olefins through a biomimetic approach.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-03-02)
Beneesh P Babu, Xu Meng, Jan-E Bäckvall
ABSTRACT
Arenes and electron-deficient olefins can be oxidatively coupled through a biomimetic Pd(OAc)2-catalyzed transformation. C-H activation of the arene partner is effected under reaction conditions of low catalyst loading, normal oxygen pressure, and using p-benzoquinone and iron phthalocyanine as electron-transfer mediators (ETMs). By controlling catalyst loading, the reaction can be made selective for either mono- or diarylation.
Product Description
Supelco
1,4-Benzoquinone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,4-Dimethoxybenzene, ReagentPlus®, 99%
Sigma-Aldrich
1,4-Dimethoxybenzene, 99%, FG
Sigma-Aldrich
p-Benzoquinone, reagent grade, ≥98%
Supelco
p-Benzoquinone, for spectrophotometric det. of amines, ≥99.5% (HPLC)