Merck
  • Superhydrophobic PLA fabrics prepared by UV photo-grafting of hydrophobic silica particles possessing vinyl groups.
Superhydrophobic PLA fabrics prepared by UV photo-grafting of hydrophobic silica particles possessing vinyl groups.
Journal of colloid and interface science (2010-02-09)
Geun Yeol Bae, Jinho Jang, Young Gyu Jeong, Won Seok Lyoo, Byung Gil Min
ABSTRACT
Superhydrophobic poly(lactic acid) (PLA) fabrics are prepared by UV photo-grafting of hydrophobic silica particles possessing vinyl functional groups on the surfaces, which is a novel one-step process to provide surface with roughness as well as hydrophobicity simultaneously. For this purpose, hydrophobic silica particles with vinyl groups and average diameter of 1.51+/-0.05 microm are synthesized via a sol-gel process. The silica particles possessing vinyl groups are found to be effectively immobilized on PLA fabrics via UV photo-grafting reaction. The water contact angle of the treated PLA fabric is measured to be approximately 150 degrees, which is high enough to exhibit the Lotus effect as a result of the superhydrophobicity.
Sigma-Aldrich
Triethoxyvinylsilane, 97%
Sigma-Aldrich
Triethoxyvinylsilane, ≥98%, deposition grade