Merck
  • Tyrosinase action on monophenols: evidence for direct enzymatic release of o-diphenol.
Tyrosinase action on monophenols: evidence for direct enzymatic release of o-diphenol.
Biochimica et biophysica acta (2001-08-22)
J N Rodríguez-López, L G Fenoll, M J Peñalver, P A García-Ruiz, R Varón, F Martínez-Ortíz, F García-Cánovas, J Tudela
ABSTRACT
Using gas chromatography-mass spectrometry, the direct enzymatic release of o-diphenol (4-tert-butylcatechol) during the action of tyrosinase on a monophenol (4-tert-butylphenol) has been demonstrated for the first time in the literature. The findings confirm the previously proposed mechanism to explain the action of tyrosinase on monophenols (J.N. Rodríguez-López, J. Tudela, R. Varón, F. García-Carmona, F. García-Cánovas, J. Biol. Chem. 267 (1992)). Oxytyrosinase, the oxidized form of the enzyme with a peroxide group, is the only form capable of catalysing the transformation of monophenols into diphenols, giving rise to an enzyme-substrate complex in the process. The o-diphenol formed is then released from the enzyme-substrate complex or oxidized to the corresponding o-quinone. In order to detect the enzymatic release of o-diphenol, the non-enzymatic evolution of the o-quinone to generate o-diphenol by weak nucleophilic attack reactions and subsequent oxidation-reduction was blocked by the nucleophilic attack of an excess of cysteine. Furthermore, the addition of catalytic quantities of an auxiliary o-diphenol (e.g. catechol) considerably increases the accumulation of 4-tert-butylcatechol. The enzyme acting on 4-tert-butylphenol generates the enzyme-4-tert-butylcatechol complex and 4-tert-butylcatechol is then released (with k(-2)) generating mettyrosinase. The auxiliary o-diphenol added (catechol) and the 4-tert-butylcatechol generated by the enzyme then enter into competition. When [catechol] > [4-tert-butylcatechol], the enzyme preferentially binds with the catechol to close the catalytic cycle, while 4-tert-butylcatechol is accumulated in the medium. In conclusion, we demonstrate that the enzyme produces 4-tert-butylcatechol from 4-tert-butylphenol, the concentration of which increases considerably in the presence of an auxiliary o-diphenol such as catechol.
Sigma-Aldrich
4-tert-Butylcatechol, SAJ first grade, ≥98.0%
Sigma-Aldrich
4-tert-Butylcatechol, ≥97.0% (HPLC)
Sigma-Aldrich
4-tert-Butylcatechol, ≥99.0%