Merck
  • Receptor transfer between immune cells by autoantibody-enhanced, CD32-driven trogocytosis is hijacked by HIV-1 to infect resting CD4 T cells.
Receptor transfer between immune cells by autoantibody-enhanced, CD32-driven trogocytosis is hijacked by HIV-1 to infect resting CD4 T cells.
Cell reports. Medicine (2024-04-06)
Manuel Albanese, Hong-Ru Chen, Madeleine Gapp, Maximilian Muenchhoff, Hsiu-Hui Yang, David Peterhoff, Katja Hoffmann, Qianhao Xiao, Adrian Ruhle, Ina Ambiel, Stephanie Schneider, Ernesto Mejías-Pérez, Marcel Stern, Paul R Wratil, Katharina Hofmann, Laura Amann, Linda Jocham, Thimo Fuchs, Alessandro F Ulivi, Simon Besson-Girard, Simon Weidlich, Jochen Schneider, Christoph D Spinner, Kathrin Sutter, Ulf Dittmer, Andreas Humpe, Philipp Baumeister, Andreas Wieser, Simon Rothenfusser, Johannes Bogner, Julia Roider, Percy Knolle, Hartmut Hengel, Ralf Wagner, Vibor Laketa, Oliver T Fackler, Oliver T Keppler
ABSTRACT
Immune cell phenotyping frequently detects lineage-unrelated receptors. Here, we report that surface receptors can be transferred from primary macrophages to CD4 T cells and identify the Fcγ receptor CD32 as driver and cargo of this trogocytotic transfer. Filamentous CD32+ nanoprotrusions deposit distinct plasma membrane patches onto target T cells. Transferred receptors confer cell migration and adhesion properties, and macrophage-derived membrane patches render resting CD4 T cells susceptible to infection by serving as hotspots for HIV-1 binding. Antibodies that recognize T cell epitopes enhance CD32-mediated trogocytosis. Such autoreactive anti-HIV-1 envelope antibodies can be found in the blood of HIV-1 patients and, consistently, the percentage of CD32+ CD4 T cells is increased in their blood. This CD32-mediated, antigen-independent cell communication mode transiently expands the receptor repertoire and functionality of immune cells. HIV-1 hijacks this mechanism by triggering the generation of trogocytosis-promoting autoantibodies to gain access to immune cells critical to its persistence.
Sigma-Aldrich
Anti-Human IgG (Fc Specific)−Agarose antibody produced in goat, affinity isolated antibody, PBS suspension
Sigma-Aldrich
Kifunensine, mannosidase inhibitor
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody