Merck
The Chick CAM as an In Vivo System to Study Stem Cell Activity.
Methods in molecular biology (Clifton, N.J.) (2022-09-27)
Marta Teixeira Pinto, Ana Sofia Ribeiro, Joana Paredes
ABSTRACT
The chick embryo chorioallantoic membrane (CAM), an extensively vascularized extraembryonic membrane, has been widely used to study several aspects of tumor development including tumor-induced angiogenesis, tumor cell proliferation, and metastasis. Based on the tumor cell/CAM system, we focused here on the identification and quantification of cancer stem cells. We validated the CAM model as a suitable model to evaluate stem cell activity in a given mixed cell population.
Sigma-Aldrich
Anti-ITGA6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution