Merck
  • Protection of K18-hACE2 mice and ferrets against SARS-CoV-2 challenge by a single-dose mucosal immunization with a parainfluenza virus 5-based COVID-19 vaccine.
Protection of K18-hACE2 mice and ferrets against SARS-CoV-2 challenge by a single-dose mucosal immunization with a parainfluenza virus 5-based COVID-19 vaccine.
Science advances (2021-07-04)
Dong An, Kun Li, Dawne K Rowe, Maria Cristina Huertas Diaz, Emily F Griffin, Ashley C Beavis, Scott K Johnson, Ian Padykula, Cheryl A Jones, Kelsey Briggs, Geng Li, Yuan Lin, Jiachen Huang, Jarrod Mousa, Melinda Brindley, Kaori Sakamoto, David K Meyerholz, Paul B McCray, S Mark Tompkins, Biao He
ABSTRACT
Transmission-blocking vaccines are urgently needed to reduce transmission of SARS-CoV 2, the cause of the COVID-19 pandemic. The upper respiratory tract is an initial site of SARS-CoV-2 infection and, for many individuals, remains the primary site of virus replication. An ideal COVID-19 vaccine should reduce upper respiratory tract virus replication and block transmission as well as protect against severe disease. Here, we optimized a vaccine candidate, parainfluenza virus 5 (PIV5) expressing the SARS-CoV-2 S protein (CVXGA1), and then demonstrated that a single-dose intranasal immunization with CVXGA1 protects against lethal infection of K18-hACE2 mice, a severe disease model. CVXGA1 immunization also prevented virus infection of ferrets and blocked contact transmission. This mucosal vaccine strategy inhibited SARS-CoV-2 replication in the upper respiratory tract, thus preventing disease progression to the lower respiratory tract. A PIV5-based mucosal vaccine provides a strategy to induce protective innate and cellular immune responses and reduce SARS-CoV-2 infection and transmission in populations.
Sigma-Aldrich
Anti-SARS-CoV-1/2 S Protein Antibody, clone hu2B3E5 ZooMAb® Chimeric Monoclonal, recombinant, expressed in HEK 293 cells
Sigma-Aldrich
SIGMAFAST OPD, tablet