Merck
  • Cortical anchoring of the microtubule cytoskeleton is essential for neuron polarity.
Cortical anchoring of the microtubule cytoskeleton is essential for neuron polarity.
eLife (2020-04-16)
Liu He, Robbelien Kooistra, Ravi Das, Ellen Oudejans, Eric van Leen, Johannes Ziegler, Sybren Portegies, Bart de Haan, Anna van Regteren Altena, Riccardo Stucchi, Af Maarten Altelaar, Stefan Wieser, Michael Krieg, Casper C Hoogenraad, Martin Harterink
ABSTRACT
The development of a polarized neuron relies on the selective transport of proteins to axons and dendrites. Although it is well known that the microtubule cytoskeleton has a central role in establishing neuronal polarity, how its specific organization is established and maintained is poorly understood. Using the in vivo model system Caenorhabditis elegans, we found that the highly conserved UNC-119 protein provides a link between the membrane-associated Ankyrin (UNC-44) and the microtubule-associated CRMP (UNC-33). Together they form a periodic membrane-associated complex that anchors axonal and dendritic microtubule bundles to the cortex. This anchoring is critical to maintain microtubule organization by opposing kinesin-1 powered microtubule sliding. Disturbing this molecular complex alters neuronal polarity and causes strong developmental defects of the nervous system leading to severely paralyzed animals.
Product Description
Roche
cOmplete ULTRA Tablets, Mini, EDTA-free, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.
Sigma-Aldrich
Nystatin Suspension, suspension, 10,000 unit/mL in DPBS, BioReagent, suitable for cell culture