Merck
  • Cis/Trans Isomerization of Unsaturated Fatty Acids in Polysorbate 80 During Light Exposure of a Monoclonal Antibody-Containing Formulation.
Cis/Trans Isomerization of Unsaturated Fatty Acids in Polysorbate 80 During Light Exposure of a Monoclonal Antibody-Containing Formulation.
Journal of pharmaceutical sciences (2019-11-13)
Indira Prajapati, Björn-Hendrik Peters, Nicholas R Larson, Yangjie Wei, Sureshkumar Choudhary, Cavan Kalonia, Suzanne Hudak, Reza Esfandiary, C Russell Middaugh, Christian Schöneich
ABSTRACT
Light exposure of a monoclonal antibody formulation containing polysorbate 80 (PS80) leads to cis/trans isomerization of monounsaturated and polyunsaturated fatty acids. This cis/trans isomerization was monitored by positive electrospray ionization mass spectrometry of intact PS80 components as well as by negative ion electrospray ionization mass spectrometry analysis of free fatty acids generated via esterase-catalyzed hydrolysis. The light-induced cis/trans isomerization of unsaturated fatty acids in PS80 required the presence of the monoclonal antibody, or, at a minimum (for mechanistic studies), a combination of N-acetyltryptophan amide and glutathione disulfide, suggesting the involvement of thiyl radicals generated by photoinduced electron transfer from Trp to the disulfide. Product analysis confirmed the conversion of PS80-bound oleic acid to elaidic acid; furthermore, together with linoleic acid, we detected conjugated linoleic acids in PS80, which underwent light-induced cis/trans isomerization.
Sigma-Aldrich
N-Acetyl-L-tryptophanamide
Sigma-Aldrich
Linoleic acid, ≥99%
Supelco
Oleic acid, analytical standard
Supelco
Elaidic acid, analytical standard