Merck
  • Neuropeptide-induced modulation of carcinogenesis in a metastatic breast cancer cell line (MDA-MB-231LUC+).
Neuropeptide-induced modulation of carcinogenesis in a metastatic breast cancer cell line (MDA-MB-231LUC+).
Cancer cell international (2019-01-02)
Silvia Gutierrez, M Danilo Boada
ABSTRACT
Metastatic cancer to bone is well-known to produce extreme pain. It has been suggested that the magnitude of this perceived pain is associated with disease progression and poor prognosis. These data suggest a potential cross-talk between cancer cells and nociceptors that contribute not only to pain, but also to cancer aggressiveness although the underlying mechanisms are yet to be stablished. The in vitro dose dependent effect of neuropeptides (NPs) (substance P [SP], calcitonin gene-related peptide and neurokinin A [NKA]) and/or its combination, on the migration and invasion of MDA-MB-231LUC+ were assessed by wound healing and collagen-based cell invasion assays, respectively. The effect of NPs on the expression of its receptors (SP [NK1] and neurokinin A receptors [NK2], CALCRL and RAMP1) and kininogen (high-molecular-weight kininogen) release to the cell culture supernatant of MDA-MB-231LUC+, were measured using western-blot analysis and an ELISA assay, respectively. Statistical significance was tested using one-way ANOVA, repeated measures ANOVA, or the paired t-test. Post-hoc testing was performed with correction for multiple comparisons as appropriate. Our data show that NPs strongly modify the chemokinetic capabilities of a cellular line commonly used as a model of metastatic cancer to bone (MDA-MB-231LUC+) and increased the expression of their receptors (NK1R, NK2R, RAMP1, and CALCRL) on these cells. Finally, we demonstrate that NPs also trigger the acute release of HMWK (Bradykinin precursor) by MDA-MB-231LUC+, a molecule with both tumorigenic and pro-nociceptive activity. Based on these observations we conclude that NPs exposure modulates this breast cancer cellular line aggressiveness by increasing its ability to migrate and invade new tissues. Furthermore, these results also support the pro nociceptive and cancer promoter role of the peripheral nervous system, during the initial stages of the disease.
Sigma-Aldrich
Anti-Substance P Receptor, from rabbit, purified by affinity chromatography
Sigma-Aldrich
QCM Collagen Cell Invasion Assay, 24-well (8 µm), Colorimetric, The Cell Invasion Assay Kit kuses a 24 well plate with 8 um pores, which is ideal for evaluation of invasive tumor cells.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse