Merck
  • A novel phosphoglucomutase-deficient mouse model reveals aberrant glycosylation and early embryonic lethality.
A novel phosphoglucomutase-deficient mouse model reveals aberrant glycosylation and early embryonic lethality.
Journal of inherited metabolic disease (2019-05-12)
Bijina Balakrishnan, Jan Verheijen, Arielle Lupo, Kimiyo Raymond, Coleman Turgeon, Yueqin Yang, Kandis L Carter, Kevin J Whitehead, Tamas Kozicz, Eva Morava, Kent Lai
ABSTRACT
Patients with phosphoglucomutase (PGM1) deficiency, a congenital disorder of glycosylation (CDG) suffer from multiple disease phenotypes. Midline cleft defects are present at birth. Overtime, additional clinical phenotypes, which include severe hypoglycemia, hepatopathy, growth retardation, hormonal deficiencies, hemostatic anomalies, frequently lethal, early-onset of dilated cardiomyopathy and myopathy emerge, reflecting the central roles of the enzyme in (glycogen) metabolism and glycosylation. To delineate the pathophysiology of the tissue-specific disease phenotypes, we constructed a constitutive Pgm2 (mouse ortholog of human PGM1)-knockout (KO) mouse model using CRISPR-Cas9 technology. After multiple crosses between heterozygous parents, we were unable to identify homozygous life births in 78 newborn pups (P = 1.59897E-06), suggesting an embryonic lethality phenotype in the homozygotes. Ultrasound studies of the course of pregnancy confirmed Pgm2-deficient pups succumb before E9.5. Oral galactose supplementation (9 mg/mL drinking water) did not rescue the lethality. Biochemical studies of tissues and skin fibroblasts harvested from heterozygous animals confirmed reduced Pgm2 enzyme activity and abundance, but no change in glycogen content. However, glycomics analyses in serum revealed an abnormal glycosylation pattern in the Pgm2+/- animals, similar to that seen in PGM1-CDG.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-PGM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1