Merck
  • An unexpected INAD PDZ tandem-mediated plcβ binding in Drosophila photo receptors.
An unexpected INAD PDZ tandem-mediated plcβ binding in Drosophila photo receptors.
eLife (2018-12-12)
Fei Ye, Yuxin Huang, Jianchao Li, Yuqian Ma, Chensu Xie, Zexu Liu, Xiaoying Deng, Jun Wan, Tian Xue, Wei Liu, Mingjie Zhang
ABSTRACT
INAD assembles key enzymes of the Drosophila compound eye photo-transduction pathway into a supramolecular complex, supporting efficient and fast light signaling. However, the molecular mechanism that governs the interaction between INAD and NORPA (phospholipase Cβ, PLCβ), a key step for the fast kinetics of the light signaling, is not known. Here, we show that the NORPA C-terminal coiled-coil domain and PDZ-binding motif (CC-PBM) synergistically bind to INAD PDZ45 tandem with an unexpected mode and unprecedented high affinity. Guided by the structure of the INAD-NORPA complex, we discover that INADL is probably a mammalian counterpart of INAD. The INADL PDZ89 tandem specifically binds to PLCβ4 with a mode that is strikingly similar to that of the INAD-NORPA complex, as revealed by the structure of the INADL PDZ89-PLCβ4 CC-PBM complex. Therefore, our study suggests that the highly specific PDZ tandem - PLCβ interactions are an evolutionarily conserved mechanism in PLCβ signaling in the animal kingdom.
Sigma-Aldrich
B834(DE3) Competent Cells - Novagen, B834 is the parental strain for BL21. These hosts are methionine auxotrophs and allow high specific activity labeling of target proteins with 35S-methionine and selenomethionine for crystallography.
Sigma-Aldrich
Rosetta(DE3) Competent Cells - Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.