Merck
Reliability of human cortical organoid generation.
Nature methods (2018-12-24)
Se-Jin Yoon, Lubayna S Elahi, Anca M Pașca, Rebecca M Marton, Aaron Gordon, Omer Revah, Yuki Miura, Elisabeth M Walczak, Gwendolyn M Holdgate, H Christina Fan, John R Huguenard, Daniel H Geschwind, Sergiu P Pașca
ABSTRACT
The differentiation of pluripotent stem cells in three-dimensional cultures can recapitulate key aspects of brain development, but protocols are prone to variable results. Here we differentiated multiple human pluripotent stem cell lines for over 100 d using our previously developed approach to generate brain-region-specific organoids called cortical spheroids and, using several assays, found that spheroid generation was highly reliable and consistent. We anticipate the use of this approach for large-scale differentiation experiments and disease modeling.
Sigma-Aldrich
Anti-Tbr1 Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)