
TSKgel® Q-STAT (7 µm) HPLC Column

Synonym(s):
TSKgel® Q-STAT Columns
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing