
Tobramycin

Synonym(s):
Nebramycin Factor 6, O-[3-Amino-3-deoxy-α-D-glucopyranosyl-(1→6)]-O-[2,6-diamino-2,3,6-trideoxy-α-D-ribohexopyranosyl-(1→4)]-2-deoxy-D-streptamine
Empirical Formula (Hill Notation):
C18H37N5O9
CAS Number:
Molecular Weight:
467.51
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing