
Testosterone enantate for system suitability

Synonym(s):
Testosterone enanthate, (17β)-17-[(1-Oxoheptyl)oxy]androst-4-en-3-one, 17β-Hydroxy-4-androsten-3-one 17-enanthate, 4-Androsten-17β-ol-3-one 17-enanthate, NSC 17591, Testosterone 17β-heptanoate
Empirical Formula (Hill Notation):
C26H40O3
CAS Number:
Molecular Weight:
400.59
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing