
Monensin sodium

Synonym(s):
Monensin sodium salt hydrate
Empirical Formula (Hill Notation):
C36H61NaO11 · xH2O
CAS Number:
Molecular Weight:
692.85 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing