Skip to Content
Merck
Saxagliptin for system suitability CRS, European Pharmacopoeia (EP) Reference Standard

Saxagliptin

Saxagliptin for system suitability CRS, European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Saxagliptin hydrate, (S)-3-Hydroxyadamantylglycine-L-cis-4,5-methanoprolinenitrile hydrate, BMS 477118, BMS 477118-11, BMS-477118, BMS-477118-11, BMS477118, BMS477118-11
Empirical Formula (Hill Notation):
C18H25N3O2H2O
CAS Number:
Molecular Weight:
333.43

Products

Brand
Feature
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade