
R-Gambogic acid

Synonym(s):
Gambogic acid, 1,5-Methano-1H,3H,11H-furo(3,4-g)pyrano(3,2-b)xanthene-1-crotonic acid, B"-Guttiferrin, Beta-Guttiferrin, Beta-Guttilactone, Cambogic acid, Guttatic acid, Guttic acid
Empirical Formula (Hill Notation):
C38H44O8
CAS Number:
Molecular Weight:
628.75
Products
Formula Weight
Manufacturer
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing