
(R)-(+)-1-Phenylethanol

Synonym(s):
(+)-Methyl phenyl carbinol, (R)-(+)-α-Methylbenzyl alcohol
Empirical Formula (Hill Notation):
C8H10O
CAS Number:
Molecular Weight:
122.16
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing