
Quinacrine mustard dihydrochloride

Synonym(s):
2-Methoxy-6-chloro-9-(4-bis[β-chloroethyl]amino-1-methylbutylamino)acridine dihydrochloride, 9-[4-(Bis(2-chloroethyl)amino)-1-methylbutylamino]-6-chloro-2-methoxyacridine dihydrochloride
Empirical Formula (Hill Notation):
C23H28Cl3N3O · 2HCl
Products
Brand
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing