
Procaine benzylpenicillin for peak identification A

Synonym(s):
Benzylpenicillin procaine salt, 3,3-Dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, compd. with 2-(diethylamino)ethyl p-aminobenzoate (1:1)
Empirical Formula (Hill Notation):
C16H18N2O4S · C13H20N2O2
CAS Number:
Molecular Weight:
570.70
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing