
Potassium pentachloronitrosylruthenate(II)

Synonym(s):
Dipotassium pentachloro(nitrosyl)ruthenate(II), di-Potassium nitrosylpentachlororuthenate(II)
Linear Formula:
K2Ru(NO)Cl5
Products
Formula Weight
Melting Point (°C)
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing