
Phenytoin sodium

Synonym(s):
5,5-Diphenylhydantoin sodium salt, 5,5-Diphenyl-2,4-imidazolidinedione, Phenytoin
Linear Formula:
C15H11N2O2Na
CAS Number:
Molecular Weight:
274.25
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing