
Pentoxifylline

Synonym(s):
3,7-Dimethyl-1-(5-oxohexyl)xanthine, Trental, Pentoxifylline
Empirical Formula (Hill Notation):
C13H18N4O3
CAS Number:
Molecular Weight:
278.31
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4