
Penicillin G sodium

Synonym(s):
Penicillin G sodium salt, Benzylpenicillin sodium salt
Empirical Formula (Hill Notation):
C16H17N2NaO4S
CAS Number:
Molecular Weight:
356.37
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2