
Parathyroid Hormone Fragment 1-34 rat

Empirical Formula (Hill Notation):
C180H291N55O48S2
CAS Number:
Molecular Weight:
4057.71
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing