
Pantoprazole Related Compound A

Synonym(s):
Pantoprazole sulfone, 5-(Difluoromethoxy)-2-{[(3,4-dimethoxy-2-pyridinyl)methyl]sulfone}-1H-benzimidazole
Empirical Formula (Hill Notation):
C16H15F2N3O5S
CAS Number:
Molecular Weight:
399.37
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2