
Palmitoyl-L-carnitine chloride

Synonym(s):
O-Palmitoyl-L-carnitine chloride
Linear Formula:
C23H46NO4 · Cl
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing