
Oxazepam Related Compound B

Synonym(s):
3-(Acetyloxy)-7-chloro-1,3-dihydro-5-phenyl-2H-1,4-benzodiazepin-2-one, 7-Chloro-1,3-dihydro-3-hydroxy-5-phenyl-2H-1,4-benzodiazepin-2-one acetate, 7-Chloro-2-oxo-5-phenyl-2,3-dihydro-1H-benzodiazepin-3-yl acetate, Oxazepam acetate
Empirical Formula (Hill Notation):
C17H13ClN2O3
CAS Number:
Molecular Weight:
328.75
Products
Brand
Feature
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2