Skip to Content
Merck
Orange G (C.I. 16230) for microscopy Certistain®

Orange G (C.I. 16230)

Orange G (C.I. 16230) for microscopy Certistain®
Synonym(s):
Orange G (C.I. 16230)
Empirical Formula (Hill Notation):
C16H10N2Na2O7S2
CAS Number:
Molecular Weight:
452.37

Products

Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing