
Okadaic acid

Synonym(s):
OA, Okadaic Acid, Prorocentrum sp.
Empirical Formula (Hill Notation):
C44H68O13
CAS Number:
Molecular Weight:
805.00
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2