
N-Hippuryl-His-Leu hydrate

Synonym(s):
N-Benzoyl-Gly-His-Leu, N-Hippuryl-L-histidyl-L-leucine hydrate
Empirical Formula (Hill Notation):
C21H27N5O5 · xH2O
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing