
N-Carbamyl-L-glutamic acid

Empirical Formula (Hill Notation):
C6H10N2O5
CAS Number:
Molecular Weight:
190.15
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Quality Segment
Application
Optical Isomer
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing