
Naproxen sodium

Synonym(s):
(S)-6-Methoxy-α-methyl-2-naphthaleneacetic acid sodium salt
Empirical Formula (Hill Notation):
C14H13NaO3
Products
Product Category
Brand
Biological Source
Feature
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4