
Monosodium glutamate

Synonym(s):
L-Glutamic acid monosodium salt monohydrate, L-2-Aminopentanedioic acid, MSG, Sodium L-glutamate
Linear Formula:
NaOOCCH2CH2CH(NH2)COOH · H2O
CAS Number:
Molecular Weight:
187.13
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing