
Meloxicam Related Compound A

Synonym(s):
Ethyl 4-hydroxy-2-methyl-1,1-dioxo-2H-1,2-benzothiazine-3-carboxylate
Empirical Formula (Hill Notation):
C12H13NO5S
CAS Number:
Molecular Weight:
283.30
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2